Gene Transcripts

Gene Symbol
Slc7a12
Entrez Gene ID
294881
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
Gene Aliases
Chromosome Location
Chr. 2: 88559677 - 88593763 on Build Rnor_6.0
UniGene ID
Rn.34357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011948.1 NP_001011948.1 1001
GenBank mRNA BC078940.1 - 1001

View Full Product Details