Gene Transcripts

Gene Symbol
Olfr94
Entrez Gene ID
258219
Gene Name
olfactory receptor 94
Gene Aliases
MOR256-32P
Chromosome Location
Chr. 17: 37196806 - 37197966 on Build GRCm38
UniGene ID
Mm.377382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011518.1 NP_001011518.1 1 67
GenBank mRNA BC139173.1 - 1 112
BC139177.1 - 1 112 8862199

View Full Product Details