Gene Transcripts

Gene Symbol
Ccdc61
Entrez Gene ID
232933
Gene Name
coiled-coil domain containing 61
Gene Aliases
C530028I08Rik, Gm159
Chromosome Location
Chr. 7: 18890883 - 18910412 on Build GRCm38
UniGene ID
Mm.333624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033314.3 NP_001028486.1 9 1340
GenBank mRNA AK082980.1 - 9 1294
AK146297.1 - 9 1341

View Full Product Details