Gene Transcripts

Gene Symbol
TSPYL2
Entrez Gene ID
64061
Gene Name
TSPY like 2
Gene Aliases
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
Chromosome Location
Chr. X: 53082327 - 53088546 on Build GRCh38
UniGene ID
Hs.136164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022117.3 NP_071400.1 4, 5 1280
XM_006724592.3 XP_006724655.1 4, 5 1313
XM_017029726.1 XP_016885215.1 3, 4 1445
XM_017029727.1 XP_016885216.1 3, 4 1445
GenBank mRNA AB015345.1 - 4, 5 520
AF254794.1 - 4, 5 1019
AF273046.1 - 4, 5 1270
AK131220.1 - 3, 4 1418
AK292462.1 - 4, 5 1273
AL713652.1 - 4, 5 1259
AY040871.1 - 4, 5 1269
BC001566.1 - 1, 2 77
BC024270.1 - 4, 5 1260 2819684

View Full Product Details