Gene Transcripts
- Gene Symbol
- C16orf70
- Entrez Gene ID
- 80262
- Gene Name
- chromosome 16 open reading frame 70
- Gene Aliases
- C16orf6, LIN10, lin-10
- Chromosome Location
- Chr. 16: 67109881 - 67148544 on Build GRCh38
- UniGene ID
- Hs.513666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320540.1 |
NP_001307469.1
|
4, 5 |
367 |
|
NM_001320541.1 |
NP_001307470.1
|
4, 5 |
408 |
|
NM_001320542.1 |
NP_001307471.1
|
4, 5 |
393 |
|
NM_025187.4 |
NP_079463.2
|
3, 4 |
423 |
|
XM_006721284.3 |
XP_006721347.1
|
4, 5 |
391 |
|
XM_011523350.2 |
XP_011521652.1
|
3, 4 |
531 |
|
XM_017023729.1 |
XP_016879218.1
|
4, 5 |
402 |
|
XM_017023730.1 |
XP_016879219.1
|
4, 5 |
405 |
|
XM_017023731.1 |
XP_016879220.1
|
4, 5 |
403 |
|
GenBank mRNA |
AF176088.1 |
- |
4, 5 |
323 |
|
AK022138.1 |
- |
2, 3 |
192 |
|
AL540304.3 |
- |
4, 5 |
302 |
|
BC004556.2 |
- |
3, 4 |
403 |
3957147 |
HY121038.1 |
- |
3, 4 |
424 |
|
HY157930.1 |
- |
4, 5 |
393 |
|
View Full Product Details