Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282405.1 | NP_001269334.1 | 3 | 1041 | |
NM_030759.4 | NP_110386.2 | 4 | 1126 | ||
XM_006717810.3 | XP_006717873.1 | 5 | 1311 | ||
GenBank mRNA | AK054957.1 | - | 4 | 1084 | |
AK301656.1 | - | 3 | 963 | ||
AL136656.1 | - | 4 | 1081 | ||
BC001345.1 | - | 1 | 710 | ||
BC011707.1 | - | 4 | 1000 | 3687848 |