Gene Transcripts
- Gene Symbol
- C6orf62
- Entrez Gene ID
- 81688
- Gene Name
- chromosome 6 open reading frame 62
- Gene Aliases
- Nbla00237, XTP12, dJ30M3.2
- Chromosome Location
- Chr. 6: 24704758 - 24720836 on Build GRCh38
- UniGene ID
- Hs.744857
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030939.4 |
NP_112201.1
|
1, 2 |
638 |
|
XM_005249433.3 |
XP_005249490.1
|
1, 2 |
1045 |
|
GenBank mRNA |
AB075502.1 |
- |
1, 2 |
625 |
|
AK022681.1 |
- |
1, 2 |
522 |
|
AL136632.2 |
- |
1, 2 |
649 |
|
AY598792.1 |
- |
1, 2 |
131 |
|
BC040260.1 |
- |
1, 2 |
150 |
|
BC047866.1 |
- |
1, 2 |
329 |
6499979 |
CR457292.1 |
- |
1, 2 |
131 |
|
CR533486.1 |
- |
1, 2 |
131 |
|
View Full Product Details