Gene Transcripts
- Gene Symbol
- LOC285696
- Entrez Gene ID
- 285696
- Gene Name
- uncharacterized LOC285696
- Gene Aliases
- Chromosome Location
- Chr. 5: 17130028 - 17217422 on Build GRCh38
- UniGene ID
- Hs.646924
- Species
- Homo sapiens
- Gene Symbol
- BASP1
- Entrez Gene ID
- 10409
- Gene Name
- brain abundant membrane attached signal protein 1
- Gene Aliases
- CAP-23, CAP23, NAP-22, NAP22
- Chromosome Location
- Chr. 5: 17216823 - 17276845 on Build GRCh38
- UniGene ID
- Hs.201641
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DA489123.1 |
- |
1 |
377 |
|
View Full Product Details