Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003213.3 | NP_003204.2 | 13 | 1589 | |
NM_201441.2 | NP_958849.1 | 12 | 1460 | ||
NM_201443.2 | NP_958851.1 | 11 | 1530 | ||
GenBank mRNA | AK222947.1 | - | 13 | 1476 | |
AY101179.1 | - | 12 | 1442 | ||
BC015497.1 | - | 13 | 1487 | 3913870 | |
BM555991.1 | - | 2 | 154 | ||
BX640921.1 | - | 11 | 1196 | ||
U63824.1 | - | 13 | 1460 |