Gene Transcripts

Gene Symbol
Shank2
Entrez Gene ID
210274
Gene Name
SH3/ankyrin domain gene 2
Gene Aliases
ProSAP1, mKIAA1022
Chromosome Location
Chr. 7: 143978056 - 144424494 on Build GRCm38
UniGene ID
Mm.483624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113373.2 NP_001106844.2 1 401
GenBank mRNA AB099695.1 - 1 28
AK139360.1 - 1 402

View Full Product Details