Gene Transcripts

Gene Symbol
Xrra1
Entrez Gene ID
446101
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
AI449753
Chromosome Location
Chr. 7: 99859218 - 99917827 on Build GRCm38
UniGene ID
Mm.440757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164258.1 NP_001157730.1 18
GenBank mRNA AK154558.1 - 1, 2 121
BC151014.1 - 18 9087927
BC151020.1 - 18 9087933

View Full Product Details