Gene Transcripts

Gene Symbol
Tmem87b
Entrez Gene ID
72477
Gene Name
transmembrane protein 87B
Gene Aliases
2610301K12Rik, 2810431I02Rik, AU014804
Chromosome Location
Chr. 2: 128818072 - 128854261 on Build GRCm38
UniGene ID
Mm.259688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028248.2 NP_082524.2 19 3059
GenBank mRNA AK050955.1 - 1 1027
AK077394.1 - 1 1334
AK161040.1 - 1 1410

View Full Product Details