Gene Transcripts
- Gene Symbol
- Npm3
- Entrez Gene ID
- 18150
- Gene Name
- nucleoplasmin 3
- Gene Aliases
- Nub1
- Chromosome Location
- Chr. 19: 45747734 - 45749563 on Build GRCm38
- UniGene ID
- Mm.1406
- Species
- Mus musculus
- Gene Symbol
- Npm3-ps1
- Entrez Gene ID
- 108176
- Gene Name
- nucleoplasmin 3, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 85076141 - 85077126 on Build GRCm38
- UniGene ID
- Mm.431982
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CK386398.1 |
- |
1 |
99 |
|
View Full Product Details