Gene Transcripts
- Gene Symbol
- Vmn2r37
- Entrez Gene ID
- 22305
- Gene Name
- vomeronasal 2, receptor 37
- Gene Aliases
- V2r14
- Chromosome Location
- Chr. 7: 9205549 - 9223653 on Build GRCm38
- UniGene ID
- Mm.302158
- Species
- Mus musculus
- Gene Symbol
- Vmn2r34
- Entrez Gene ID
- 100042636
- Gene Name
- vomeronasal 2, receptor 34
- Gene Aliases
- ENSMUSG00000070841
- Chromosome Location
- Chr. 7: 7671829 - 7689398 on Build GRCm38
- UniGene ID
- Mm.302158
- Species
- Mus musculus
- Gene Symbol
- Vmn2r45
- Entrez Gene ID
- 100042810
- Gene Name
- vomeronasal 2, receptor 45
- Gene Aliases
- Chromosome Location
- Chr. 7: 8471469 - 8488959 on Build GRCm38
- UniGene ID
- Mm.469890
- Species
- Mus musculus
View Full Product Details