Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083959.1 | NP_001077428.1 | 5 | 510 | |
NM_001083960.1 | NP_001077429.1 | 5 | 471 | ||
NM_012046.2 | NP_036176.1 | 6 | 585 | ||
GenBank mRNA | AF126400.1 | - | 5 | 510 | |
AF149309.1 | - | 5 | 434 | ||
AF163053.1 | - | 5 | 471 | ||
AF163054.1 | - | 5 | 471 | ||
AF165313.1 | - | 5 | 464 | ||
AF167439.1 | - | 5 | 503 | ||
AF169386.1 | - | 6 | 582 | ||
AF173848.1 | - | 6 | 578 | ||
AK088498.1 | - | 3 | 1644 |