Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026160.4 | NP_080436.1 | 4 | 1616 | |
GenBank mRNA | AK003106.1 | - | 4 | 1615 | |
AK132329.1 | - | 1625 | |||
AK172322.1 | - | 1 | 1175 | ||
BC068180.1 | - | 4 | 1619 | 30634082 | |
BC086774.1 | - | 3 | 4043 | ||
BC110308.1 | - | 4 | 1604 | 5319360 |