Gene Transcripts
- Gene Symbol
- Xpnpep2
- Entrez Gene ID
- 170745
- Gene Name
- X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
- Gene Aliases
- 9030008G12Rik, mAPP
- Chromosome Location
- Chr. X: 48106913 - 48136982 on Build GRCm38
- UniGene ID
- Mm.129279
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289729.1 |
NP_001276658.1
|
5, 6 |
828 |
|
NM_133213.2 |
NP_573476.2
|
5, 6 |
828 |
|
GenBank mRNA |
AF367247.1 |
- |
5, 6 |
492 |
|
AF428102.1 |
- |
5, 6 |
679 |
|
AK078835.1 |
- |
5, 6 |
635 |
|
AK085520.1 |
- |
5, 6 |
626 |
|
AK087627.1 |
- |
5, 6 |
635 |
|
AK164538.1 |
- |
5, 6 |
656 |
|
AK172086.1 |
- |
5, 6 |
829 |
|
BC140977.1 |
- |
5, 6 |
472 |
9055589 |
View Full Product Details