Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172632.2 | NP_766220.2 | 2 | 1093 | |
GenBank mRNA | AK079654.1 | - | 2 | 1071 | |
AK138531.1 | - | 1 | 1162 | ||
AK157930.1 | - | 2 | 1033 | ||
AK162916.1 | - | 1 | 1162 | ||
BC058942.1 | - | 1 | 496 | 6821544 | |
BC062911.1 | - | 1 | 496 | 6821544 | |
BC094564.1 | - | 2 | 1452 |