Gene Transcripts

Gene Symbol
Arl15
Entrez Gene ID
218639
Gene Name
ADP-ribosylation factor-like 15
Gene Aliases
A430036I03, Arfrp2, C230032K13Rik
Chromosome Location
Chr. 13: 113794506 - 114157462 on Build GRCm38
UniGene ID
Mm.79837
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172595.4 NP_766183.1 5 2406
GenBank mRNA AK039965.1 - 5 2370
AK048736.1 - 5 2291
AK050979.1 - 5 2321
AK052164.1 - 5 2358

View Full Product Details