Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000956.3 | NP_000947.2 | 1 | 1043 | |
GenBank mRNA | AY275471.1 | - | 1 | 792 | |
BC093927.1 | - | 1 | 845 | 7939772 | |
BC093929.1 | - | 1 | 845 | 7939774 | |
U19487.1 | - | 1 | 948 | ||
X83868.1 | - | 1 | 792 |