Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
Chromosome Location
Chr. 1: 145911348 - 145918924 on Build GRCh38
UniGene ID
Hs.714608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184795.1 NP_001171724.1 4 668
NM_003846.2 NP_003837.1 4 862
NR_073491.1 4 858
NR_073492.1 4 852
NR_073493.2 4 1074
GenBank mRNA AB018080.1 - 4 644
AF093670.1 - 4 642
AK023991.1 - 4 626
AK094173.1 - 1 1462
AK301983.1 - 4 628
BC011963.1 - 4 642 3866690
CR542047.1 - 4 626

View Full Product Details