Gene Transcripts
- Gene Symbol
- ATP6AP2
- Entrez Gene ID
- 10159
- Gene Name
- ATPase H+ transporting accessory protein 2
- Gene Aliases
- APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
- Chromosome Location
- Chr. X: 40580964 - 40606637 on Build GRCh38
- UniGene ID
- Hs.495960
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005765.2 |
NP_005756.2
|
3, 4 |
399 |
|
GenBank mRNA |
AF109363.1 |
- |
3, 4 |
360 |
|
AF248966.1 |
- |
3, 4 |
404 |
|
AF291814.1 |
- |
3, 4 |
386 |
|
AK075382.1 |
- |
3, 4 |
457 |
|
AK296614.1 |
- |
2, 3 |
262 |
|
AK297007.1 |
- |
3, 4 |
370 |
|
AK314633.1 |
- |
3, 4 |
350 |
|
AL049929.1 |
- |
2, 3 |
239 |
|
AY038990.1 |
- |
3, 4 |
402 |
|
AY429341.1 |
- |
2, 3 |
262 |
|
BC084541.1 |
- |
3, 4 |
379 |
6575732 |
View Full Product Details