Gene Transcripts

Gene Symbol
SHANK2
Entrez Gene ID
22941
Gene Name
SH3 and multiple ankyrin repeat domains 2
Gene Aliases
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
Chromosome Location
Chr. 11: 70467856 - 71252724 on Build GRCh38
UniGene ID
Hs.268726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133266.4 NP_573573.2 1 117
NR_110766.1 1 117
GenBank mRNA AB208027.1 - 1 95
BC093885.1 - 1 117
BC112097.1 - 1 117
BC113876.1 - 1 40
BC114484.1 - 1 39
BC143389.1 - 1 117

View Full Product Details