Gene Transcripts

Gene Symbol
MPC1
Entrez Gene ID
51660
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L, CGI-129, MPYCD, dJ68L15.3
Chromosome Location
Chr. 6: 166364919 - 166383013 on Build GRCh38
UniGene ID
Hs.172755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270879.1 NP_001257808.1 4, 5 536
NM_016098.3 NP_057182.1 3, 4 320
NR_073093.1 4, 5 549
XM_011535894.1 XP_011534196.1 3, 4 1319
XM_011535895.1 XP_011534197.1 2, 3 1103
GenBank mRNA AF125101.1 - 3, 4 305
AF151887.1 - 3, 4 302
AF275811.1 - 1, 2 27
AK312201.1 - 3, 4 244
AL556427.3 - 3, 4 239
AL561589.3 - 4, 5 424
BC000810.1 - 3, 4 230 3452973
BF665465.1 - 4, 5 408
BG828174.1 - 3, 4 278

View Full Product Details