Gene Transcripts
- Gene Symbol
- PLEKHB1
- Entrez Gene ID
- 58473
- Gene Name
- pleckstrin homology domain containing B1
- Gene Aliases
- KPL1, PHR1, PHRET1
- Chromosome Location
- Chr. 11: 73646178 - 73662819 on Build GRCh38
- UniGene ID
- Hs.445489
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130033.1 |
NP_001123505.1
|
4, 5 |
776 |
|
NM_001130034.1 |
NP_001123506.1
|
4, 5 |
517 |
|
NM_001130035.1 |
NP_001123507.1
|
4, 5 |
517 |
|
NM_001130036.1 |
NP_001123508.1
|
3, 4 |
649 |
|
NM_021200.2 |
NP_067023.1
|
4, 5 |
776 |
|
XM_011545192.2 |
XP_011543494.1
|
4, 5 |
1350 |
|
XM_011545193.2 |
XP_011543495.1
|
4, 5 |
1350 |
|
GenBank mRNA |
AB209716.1 |
- |
4, 5 |
429 |
|
AF081583.1 |
- |
3, 4 |
293 |
|
AF093249.1 |
- |
4, 5 |
401 |
|
AF100612.1 |
- |
4, 5 |
401 |
|
AF101054.1 |
- |
4, 5 |
772 |
|
AK289620.1 |
- |
4, 5 |
429 |
|
AK289909.1 |
- |
4, 5 |
395 |
|
AK290109.1 |
- |
3, 4 |
649 |
|
AK296033.1 |
- |
4, 5 |
411 |
|
AK301299.1 |
- |
4, 5 |
412 |
|
AK311158.1 |
- |
5, 6 |
274 |
|
AK314748.1 |
- |
4, 5 |
373 |
|
BC008075.2 |
- |
4, 5 |
455 |
3050369 |
DC305763.1 |
- |
4, 5 |
517 |
|
U89715.1 |
- |
4, 5 |
772 |
|
View Full Product Details