Gene Transcripts

Gene Symbol
PLK5
Entrez Gene ID
126520
Gene Name
polo like kinase 5
Gene Aliases
PLK-5, PLK5P, SgK384ps
Chromosome Location
Chr. 19: 1524074 - 1535456 on Build GRCh38
UniGene ID
Hs.195352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243079.1 NP_001230008.1 14 1709
GenBank mRNA AK054808.1 - 14 1709
AK122648.1 - 13 1881
BC100791.2 - 11 1273
BC100792.1 - 11 1273
DQ424898.1 - 7 879

View Full Product Details