Gene Transcripts

Gene Symbol
SHANK2
Entrez Gene ID
22941
Gene Name
SH3 and multiple ankyrin repeat domains 2
Gene Aliases
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
Chromosome Location
Chr. 11: 70467856 - 71252724 on Build GRCh38
UniGene ID
Hs.268726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012309.4 NP_036441.2 9 1153
XM_017017387.1 XP_016872876.1 10 1258
XM_017017388.1 XP_016872877.1 10 1280
XM_017017389.1 XP_016872878.1 10 1453
GenBank mRNA AB208025.1 - 9 1119
AK096045.1 - 12 1330
DQ152234.1 - 8 1041

View Full Product Details