Gene Transcripts

Gene Symbol
CATSPER1
Entrez Gene ID
117144
Gene Name
cation channel sperm associated 1
Gene Aliases
CATSPER, SPGF7
Chromosome Location
Chr. 11: 66016752 - 66026518 on Build GRCh38
UniGene ID
Hs.189105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053054.3 NP_444282.3 6 1988
GenBank mRNA AF407333.1 - 6 1850
BC032950.2 - 6 1987 4822261
BC036522.2 - 6 1950 5268567

View Full Product Details