Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304485.1 | NP_001291414.1 | 5 | 592 | |
NM_001304486.1 | NP_001291415.1 | 4 | 470 | ||
NM_001304487.1 | NP_001291416.1 | 4 | 504 | ||
NM_001304488.1 | NP_001291417.1 | 5 | 489 | ||
NM_001304489.1 | NP_001291418.1 | 5 | 474 | ||
NM_001304490.1 | NP_001291419.1 | 5 | 471 | ||
NM_001304491.1 | NP_001291420.1 | 4 | 383 | ||
NM_012460.3 | NP_036592.1 | 6 | 781 | ||
NR_130750.1 | 5 | 716 | |||
NR_130751.1 | 4 | 406 | |||
NR_130752.1 | 3 | 405 | |||
NR_130753.1 | 3 | 397 | |||
GenBank mRNA | AF150100.1 | - | 5 | 472 | |
AF152353.1 | - | 3 | 255 | ||
AK000666.1 | - | 5 | 490 | ||
AK312095.1 | - | 4 | 416 | ||
BC020213.1 | - | 6 | 681 | 4640518 | |
BC054875.1 | - | 4 | 394 | 6663143 | |
BF034731.1 | - | 5 | 436 | ||
BG249379.1 | - | 5 | 361 | ||
BM811095.1 | - | 5 | 628 | ||
DB110327.1 | - | 3 | 334 | ||
DN912572.1 | - | 5 | 403 | ||
HY041857.1 | - | 4 | 286 | ||
HY096567.1 | - | 4 | 335 | ||
W74224.1 | - | 5 | 377 |