Gene Transcripts

Gene Symbol
SPATA1
Entrez Gene ID
100505741
Gene Name
spermatogenesis associated 1
Gene Aliases
SP-2, SPAP1
Chromosome Location
Chr. 1: 84506301 - 84556495 on Build GRCh38
UniGene ID
Hs.584956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310156.1 NP_001297085.1 11, 12 1380
XM_011542514.2 XP_011540816.1 10, 11 1254
GenBank mRNA AF306347.1 - 11, 12 1348

View Full Product Details