Gene Transcripts

Gene Symbol
Akr1c18
Entrez Gene ID
105349
Gene Name
aldo-keto reductase family 1, member C18
Gene Aliases
AW146047
Chromosome Location
Chr. 13: 4132615 - 4150651 on Build GRCm38
UniGene ID
Mm.41337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134066.2 NP_598827.1 6 695
GenBank mRNA AB059565.1 - 6 695
AK153898.1 - 6 713
BC034259.1 - 5 614 4238589

View Full Product Details