Gene Transcripts

Gene Symbol
Aspa
Entrez Gene ID
11484
Gene Name
aspartoacylase
Gene Aliases
Acy-2, Acy2, nur7
Chromosome Location
Chr. 11: 73304988 - 73326868 on Build GRCm38
UniGene ID
Mm.293574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023113.5 NP_075602.2 5 835
GenBank mRNA AF212998.1 - 5 685
AK002859.1 - 5 833
AK008354.1 - 5 838
BC024934.1 - 5 799 4012726

View Full Product Details