Gene Transcripts

Gene Symbol
Ufsp2
Entrez Gene ID
192169
Gene Name
UFM1-specific peptidase 2
Gene Aliases
1810047C23Rik
Chromosome Location
Chr. 8: 45975528 - 45996958 on Build GRCm38
UniGene ID
Mm.210882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138668.2 NP_619609.1 3 199
GenBank mRNA AK075795.1 - 3 194
AK078309.1 - 3 200
AK160164.1 - 3 194
BC005503.1 - 3 200 3490744

View Full Product Details