Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008964.4 | NP_032990.1 | 2 | 1847 | |
GenBank mRNA | AK036531.1 | - | 2 | 1765 | |
AK054132.1 | - | 2 | 1368 | ||
AK086904.1 | - | 2 | 1762 | ||
AK136577.1 | - | 2 | 860 | ||
AK137515.1 | - | 2 | 1057 | ||
BC005440.1 | - | 2 | 1355 | 3487087 | |
D50589.1 | - | 2 | 1138 |