Gene Transcripts

Gene Symbol
Rundc3b
Entrez Gene ID
242819
Gene Name
RUN domain containing 3B
Gene Aliases
Gm440
Chromosome Location
Chr. 5: 8490336 - 8623027 on Build GRCm38
UniGene ID
Mm.332192
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198620.1 NP_941022.1 4 730
GenBank mRNA BC058808.1 - 4 730

View Full Product Details