Gene Transcripts

Gene Symbol
Tinag
Entrez Gene ID
26944
Gene Name
tubulointerstitial nephritis antigen
Gene Aliases
AI452335, TIN-ag
Chromosome Location
Chr. 9: 76951687 - 77045806 on Build GRCm38
UniGene ID
Mm.23199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012033.3 NP_036163.3 11 1489
GenBank mRNA AF153366.1 - 11 1431
AK008374.1 - 11 1465
BC010745.1 - 11 1434 4239192

View Full Product Details