Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009818.1 | NP_001009818.1 | 4 | 561 | |
GenBank mRNA | AK028475.1 | - | 4 | 493 | |
AK136063.1 | - | 2 | 190 | ||
AK166517.1 | - | 4 | 582 | ||
AK170945.1 | - | 3 | 371 | ||
AK171368.1 | - | 4 | 582 | ||
BC031456.1 | - | 4 | 468 | 4037515 | |
BC062206.1 | - | 4 | 602 | 5150858 | |
BC064466.1 | - | 4 | 561 | 6825549 |