Gene Transcripts

Gene Symbol
Lap3
Entrez Gene ID
66988
Gene Name
leucine aminopeptidase 3
Gene Aliases
2410015L10Rik, AA410100, LAP-3, Lap, Lapep, Pep-7, Pep-S, Pep7, Peps
Chromosome Location
Chr. 5: 45493374 - 45512691 on Build GRCm38
UniGene ID
Mm.286830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024434.6 NP_077754.3 3 344
GenBank mRNA AF334160.1 - 3 305
AK002819.1 - 3 280
AK005334.1 - 3 257
AK008600.1 - 3 231
AK010384.1 - 3 230
AK010502.1 - 3 297
AK011604.1 - 3 230
AK089023.1 - 3 311
AK166958.1 - 3 228
AK169032.1 - 3 230
BC016536.1 - 3 344 3488469
CT010237.1 - 3 258
CT010301.1 - 3 258

View Full Product Details