Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025959.3 | NP_080235.2 | 5 | 313 | |
GenBank mRNA | AK008996.1 | - | 57 | ||
AK012174.1 | - | 5 | 314 | ||
AK014354.1 | - | 5 | 301 | ||
AK144728.1 | - | 310 | |||
AK166961.1 | - | 5 | 298 | ||
BC037099.1 | - | 5 | 311 | ||
BC043044.1 | - | 292 | |||
BC057997.1 | - | 269 | |||
BC116752.1 | - | 5 | 313 | 40126071 | |
BC119168.1 | - | 5 | 313 | 8733917 |