Gene Transcripts

Gene Symbol
Tmem19
Entrez Gene ID
67226
Gene Name
transmembrane protein 19
Gene Aliases
2810428F02Rik, AW125171
Chromosome Location
Chr. 10: 115340739 - 115362524 on Build GRCm38
UniGene ID
Mm.29647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133683.3 NP_598444.1 2 292
GenBank mRNA AK018383.1 - 2 241
AK034298.1 - 2 298
AK036228.1 - 1 369
AK042426.1 - 1 347
AK136778.1 - 281
AK138931.1 - 2 280
AK155275.1 - 2 253
AK170143.1 - 2 252
AK171087.1 - 2 252
AK209903.1 - 1 38
AK213890.1 - 1 38
BC016895.1 - 2 294 4239007

View Full Product Details