Gene Transcripts
- Gene Symbol
- Ttc29
- Entrez Gene ID
- 73301
- Gene Name
- tetratricopeptide repeat domain 29
- Gene Aliases
- 1700031F13Rik
- Chromosome Location
- Chr. 8: 78213302 - 78394326 on Build GRCm38
- UniGene ID
- Mm.61243
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_183096.3 |
NP_898919.3
|
12 |
1690 |
|
GenBank mRNA |
BC048386.1 |
- |
12 |
1690 |
|
View Full Product Details