Gene Transcripts

Gene Symbol
Ssna1
Entrez Gene ID
68475
Gene Name
Sjogren's syndrome nuclear autoantigen 1
Gene Aliases
1110003H09Rik, 1190004J23Rik, AA934184, N14, NA14
Chromosome Location
Chr. 2: 25271039 - 25272418 on Build GRCm38
UniGene ID
Mm.21444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023464.2 NP_075953.1 3 346
GenBank mRNA AB041656.1 - 3 340
AK003356.2 - 3 347
AK004482.1 - 1 225
AK088418.1 - 3 278
AK158412.1 - 463
AK208309.1 - 2 38
BC023033.1 - 3 309
BC037081.1 - 3 323 5098653
BC048360.1 - 3 324 5067090
BY703613.1 - 3 347

View Full Product Details