Gene Transcripts

Gene Symbol
Ppp2r3d
Entrez Gene ID
19054
Gene Name
protein phosphatase 2 (formerly 2A), regulatory subunit B'', delta
Gene Aliases
PP2A B''-PR59, PP2A PR59, PR59, Ppp2r3, Ppp2r3a
Chromosome Location
Chr. 9: 124438769 - 124441010 on Build GRCm38
UniGene ID
Mm.271249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163415.1 NP_001156887.1 1687
NM_011154.2 NP_035284.2 1453
GenBank mRNA AF050165.1 - 1450
AK054122.1 - 1795
AK143212.1 - 1730
BC027415.1 - 1430 4235453
BC059852.1 - 1718 6402777
BC095938.1 - 577
BC096544.1 - 1133 6834351

View Full Product Details