Gene Transcripts

Gene Symbol
Vat1
Entrez Gene ID
26949
Gene Name
vesicle amine transport protein 1 homolog (T californica)
Gene Aliases
VAT-1
Chromosome Location
Chr. 11: 101458748 - 101466199 on Build GRCm38
UniGene ID
Mm.29210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012037.2 NP_036167.1 3 780
GenBank mRNA AK034795.1 - 3 782
AK145193.1 - 3 814
AK154178.1 - 765
AK154965.1 - 3 763
AK155342.1 - 3 781
AK159310.1 - 3 723
AK159315.1 - 3 765
AK160891.1 - 3 772
AK170371.1 - 3 761
AK178348.1 - 1 34
AK183725.1 - 1 34
AK185056.1 - 2 114
AK199516.1 - 1 34
BC025496.1 - 3 736 5320239
BC036169.1 - 2 222
BC085504.1 - 3 699
BC099678.1 - 3 762 6311681
X95562.1 - 3 402

View Full Product Details