Gene Transcripts

Gene Symbol
Ttyh1
Entrez Gene ID
292597
Gene Name
tweety family member 1
Gene Aliases
Chromosome Location
Chr. 1: 73734565 - 73753157 on Build Rnor_6.0
UniGene ID
Rn.14889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106225.1 NP_001099695.1 10 1153
XM_006228216.2 XP_006228278.1 10 1177
GenBank mRNA BC168158.1 - 10 1148
FQ212233.1 - 7 618

View Full Product Details