Gene Transcripts

Gene Symbol
Poldip3
Entrez Gene ID
315170
Gene Name
polymerase (DNA-directed), delta interacting protein 3
Gene Aliases
Chromosome Location
Chr. 7: 123992167 - 124020596 on Build Rnor_6.0
UniGene ID
Rn.3893
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130506.1 NP_001123978.1 2 203
XM_006242090.2 XP_006242152.1 2 225

View Full Product Details