Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191586.1 | NP_001178515.1 | 14 | 5172 | |
XM_006256659.2 | XP_006256721.1 | 14 | 5132 | ||
XM_006256660.2 | XP_006256722.1 | 14 | 5202 | ||
XM_006256661.2 | XP_006256723.1 | 14 | 5296 | ||
XM_006256663.2 | XP_006256725.1 | 13 | 5186 | ||
XM_006256664.2 | XP_006256726.1 | 14 | 5138 | ||
XM_006256665.2 | XP_006256727.1 | 13 | 5084 | ||
XM_008773074.1 | XP_008771296.1 | 14 | 10740 |