Gene Transcripts

Gene Symbol
Map7d2
Entrez Gene ID
317508
Gene Name
MAP7 domain containing 2
Gene Aliases
Brelin, Mtap7d2, RGD1564852
Chromosome Location
Chr. X: 38400578 - 38467825 on Build Rnor_6.0
UniGene ID
Rn.231886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289778.1 NP_001276707.1 8 1135
GenBank mRNA AB266744.1 - 8 1135

View Full Product Details