Gene Transcripts

Gene Symbol
Slc35d3
Entrez Gene ID
308717
Gene Name
solute carrier family 35, member D3
Gene Aliases
Chromosome Location
Chr. 1: 15298526 - 15301998 on Build Rnor_6.0
UniGene ID
Rn.110407
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107522.1 NP_001100992.1 2 1300

View Full Product Details