Gene Transcripts

Gene Symbol
Vom2r10
Entrez Gene ID
292414
Gene Name
vomeronasal 2 receptor, 10
Gene Aliases
RGD1565464
Chromosome Location
Chr. 1: 86103298 - 86149320 on Build Rnor_6.0
UniGene ID
Rn.217941
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099469.1 NP_001092939.1 6 2474

View Full Product Details